Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ly49si2
Entrez Gene ID
494207
Gene Name
immunoreceptor Ly49si2
Gene Aliases
-
Chromosome Location
Chr.4: 163920579 - 163954830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009498.1 NP_001009498.1 - 673 101
XM_008763406.1 XP_008761628.1 - 686 101
XM_008763407.1 XP_008761629.1 - 686 101
GenBank mRNA AY659933.1 - - 673 101

Gene Symbol
LOC690097
Entrez Gene ID
690097
Gene Name
similar to immunoreceptor Ly49si3
Gene Aliases
-
Chromosome Location
Chr.4: 164027995 - 164053891 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763433.1 XP_008761655.1 6 - 7 1099 101

Gene Symbol
Ly49si3
Entrez Gene ID
494209
Gene Name
immunoreceptor Ly49si3
Gene Aliases
-
Chromosome Location
Chr.4: 163986547 - 164028470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009919.1 NP_001009919.1 5 - 6 673 101
XM_006237116.2 XP_006237178.1 - 767 101
XM_008763410.1 XP_008761632.1 6 - 7 954 101
XM_008763411.1 XP_008761633.1 6 - 7 1591 101
XM_008763412.1 XP_008761634.1 6 - 7 748 101
XM_008763413.1 XP_008761635.1 6 - 7 954 101
GenBank mRNA AY653730.1 - 5 - 6 673 101

View Full Product Details