Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nfkbil1
Entrez Gene ID
361794
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Aliases
-
Chromosome Location
Chr.20: 4823612 - 4839202 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212509.2 NP_997674.1 1 - 2 168 100
GenBank mRNA BC099772.1 - 1 - 2 112 100

View Full Product Details