Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmsb10
Entrez Gene ID
50665
Gene Name
thymosin, beta 10
Gene Aliases
Ptmb10, THYb10
Chromosome Location
Chr.4: 100882216 - 100883294 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021261.2 NP_067084.1 1 - 2 64 74
GenBank mRNA CF977046.1 - 1 - 2 68 74
FQ221466.1 - 1 - 2 62 74
FQ223015.1 - 1 - 2 62 74
FQ224120.1 - 1 - 2 64 74
FQ224544.1 - 1 - 2 62 74
FQ229044.1 - 1 - 2 63 74

View Full Product Details