Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpg1
Entrez Gene ID
299247
Gene Name
candidate plasticity gene 1
Gene Aliases
-
Chromosome Location
Chr.6: 123034304 - 123046297 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178104.2 NP_835205.1 1 - 2 293 108
GenBank mRNA X73292.1 - 1 - 2 293 108

View Full Product Details