Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom1r81
Entrez Gene ID
494313
Gene Name
vomeronasal 1 receptor 81
Gene Aliases
V1rc2
Chromosome Location
Chr.4: 88048267 - 88049199 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008970.1 NP_001008970.1 1 - 1 349 95
GenBank mRNA AY510288.1 - 1 - 1 349 95

View Full Product Details