Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ly49si3
Entrez Gene ID
494209
Gene Name
immunoreceptor Ly49si3
Gene Aliases
-
Chromosome Location
Chr.4: 163986547 - 164028470 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009919.1 NP_001009919.1 3 - 4 418 100
XM_006237116.2 XP_006237178.1 - 100
XM_008763410.1 XP_008761632.1 4 - 5 100
XM_008763411.1 XP_008761633.1 4 - 5 100
XM_008763412.1 XP_008761634.1 4 - 5 100
XM_008763413.1 XP_008761635.1 4 - 5 100
XM_008763414.1 XP_008761636.1 4 - 5 100
XM_008763415.1 XP_008761637.1 4 - 5 100
GenBank mRNA AY653730.1 - 3 - 4 418 100

Gene Symbol
LOC100910264
Entrez Gene ID
100910264
Gene Name
T-cell surface glycoprotein YE1/48-like
Gene Aliases
-
Chromosome Location
Chr.4: 163804762 - 163812982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008763346.1 XP_008761568.1 3 - 4 100

Gene Symbol
Ly49si2
Entrez Gene ID
494207
Gene Name
immunoreceptor Ly49si2
Gene Aliases
-
Chromosome Location
Chr.4: 163920579 - 163954830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009498.1 NP_001009498.1 - 100
XM_008763406.1 XP_008761628.1 - 100
XM_008763407.1 XP_008761629.1 - 100
XM_008763408.1 XP_008761630.1 - 100
XM_008763409.1 XP_008761631.1 - 100
GenBank mRNA AY659933.1 - - 100

View Full Product Details