Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Spetex-2C
- Entrez Gene ID
-
364255
- Gene Name
- Spetex-2C protein
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- Spetex-2D
- Entrez Gene ID
-
364260
- Gene Name
- Spetex-2D protein
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- Spetex-2E
- Entrez Gene ID
-
361012
- Gene Name
- Spetex-2E protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 5414416 - 5417642 on Build Rnor_6.0
- Gene Symbol
- Spetex-2F
- Entrez Gene ID
-
316940
- Gene Name
- Spetex-2F protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 5463852 - 5509839 on Build Rnor_6.0
- Gene Symbol
- Spetex-2G
- Entrez Gene ID
-
361099
- Gene Name
- Spetex-2G protein
- Gene Aliases
-
-
- Chromosome Location
-
-
- Gene Symbol
- Spetex-2H
- Entrez Gene ID
-
361008
- Gene Name
- Spetex-2H protein
- Gene Aliases
-
Spergen-2H
- Chromosome Location
-
-
View Full Product Details