The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034950.2 | NP_001030122.2 | 3 - 4 | 383 | 118 |
GenBank mRNA | AF198441.1 | - | 3 - 4 | 355 | 118 |
AY327506.1 | - | 3 - 4 | 383 | 118 |