Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1ql3
Entrez Gene ID
680404
Gene Name
complement component 1, q subcomponent-like 3
Gene Aliases
-
Chromosome Location
Chr.17: 80311698 - 80332568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109403.2 NP_001102873.1 1 - 2 589 65
XM_008771889.1 XP_008770111.1 3 - 4 5048 65
XM_008771890.1 XP_008770112.1 4 - 5 8106 65
XM_008771891.1 XP_008770113.1 3 - 4 4371 65

View Full Product Details