Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ghdc
Entrez Gene ID
303542
Gene Name
GH3 domain containing
Gene Aliases
RGD1308881
Chromosome Location
Chr.10: 88672834 - 88677055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191651.1 NP_001178580.1 3 - 4 469 83

View Full Product Details