Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfatc1
Entrez Gene ID
100361818
Gene Name
nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Gene Aliases
-
Chromosome Location
Chr.18: 77203517 - 77322690 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244933.1 NP_001231862.1 9 - 10 2785 68
GenBank mRNA FQ222003.1 - 9 - 10 2785 68
FQ223020.1 - 9 - 10 2785 68

View Full Product Details