Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pop5
Entrez Gene ID
117241
Gene Name
processing of precursor 5, ribonuclease P/MRP subunit (S. cerevisiae)
Gene Aliases
Rnasep3, rpP2
Chromosome Location
Chr.12: 47138346 - 47142852 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105752.2 NP_001099222.2 3 - 4 837 69

View Full Product Details