Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmd6
Entrez Gene ID
257646
Gene Name
FERM domain containing 6
Gene Aliases
-
Chromosome Location
Chr.6: 93281382 - 93358888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271054.1 NP_001257983.1 12 - 13 1728 70
XM_006240151.2 XP_006240213.1 12 - 13 1778 70
XM_006240152.2 XP_006240214.1 13 - 14 1858 70
XM_006240153.2 XP_006240215.1 12 - 13 2202 70

View Full Product Details