The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021595.2 | NP_067606.1 | 3 - 4 | 578 | 99 |
XM_008763255.1 | XP_008761477.1 | 3 - 4 | 1088 | 99 | |
XM_008763256.1 | XP_008761478.1 | 4 - 5 | 1005 | 99 | |
XM_008763257.1 | XP_008761479.1 | 4 - 5 | 764 | 99 | |
XM_008763258.1 | XP_008761480.1 | 4 - 5 | 915 | 99 | |
XM_008763259.1 | XP_008761481.1 | 3 - 4 | 724 | 99 | |
GenBank mRNA | AB040815.1 | - | 3 - 4 | 579 | 99 |
AF250322.1 | - | 3 - 4 | 579 | 99 |