Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ftl1
Entrez Gene ID
29292
Gene Name
ferritin light chain 1
Gene Aliases
Ftl
Chromosome Location
Chr.1: 101448190 - 101450034 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022500.4 NP_071945.3 1 - 2 307 76
GenBank mRNA BC061525.1 - 1 - 2 311 76
BC086583.1 - 1 - 2 288 76
BC088756.1 - 1 - 2 288 76
FQ209407.1 - 1 - 2 307 76
FQ209694.1 - 1 - 2 307 76
FQ209816.1 - 1 - 2 307 76
FQ218577.1 - 1 - 2 307 76
FQ219483.1 - 1 - 2 307 76
FQ219685.1 - 1 - 2 307 76
FQ219809.1 - 1 - 2 307 76
FQ219930.1 - 1 - 2 306 76
FQ220095.1 - 1 - 2 307 76
FQ220123.1 - 1 - 2 307 76
FQ220651.1 - 1 - 2 307 76
FQ220814.1 - 1 - 2 307 76
FQ220884.1 - 1 - 2 307 76
FQ221898.1 - 1 - 2 307 76
FQ223227.1 - 1 - 2 307 76
FQ226019.1 - 1 - 2 307 76
FQ226254.1 - 1 - 2 263 76
FQ226458.1 - 1 - 2 306 76
FQ227401.1 - 1 - 2 307 76
FQ227800.1 - 1 - 2 307 76
FQ227843.1 - 1 - 2 307 76
FQ228050.1 - 1 - 2 307 76
FQ228585.1 - 1 - 2 307 76
FQ229176.1 - 1 - 2 307 76
FQ229284.1 - 1 - 2 307 76
FQ229370.1 - 1 - 2 307 76
FQ229673.1 - 1 - 2 305 76
FQ230098.1 - 1 - 2 305 76
FQ230445.1 - 1 - 2 307 76
FQ230659.1 - 1 - 2 303 76
FQ231322.1 - 1 - 2 307 76
FQ231373.1 - 1 - 2 307 76
FQ231464.1 - 1 - 2 307 76
FQ231570.1 - 1 - 2 307 76
FQ231615.1 - 1 - 2 307 76
FQ231781.1 - 1 - 2 303 76
FQ231957.1 - 1 - 2 307 76
FQ232031.1 - 1 - 2 307 76
FQ232167.1 - 1 - 2 307 76
FQ232517.1 - 1 - 2 307 76
FQ233772.1 - 1 - 2 307 76
FQ233996.1 - 1 - 2 307 76
FQ234074.1 - 1 - 2 307 76
FQ234603.1 - 1 - 2 307 76
FQ234647.1 - 1 - 2 307 76
FQ235044.1 - 1 - 2 307 76
FQ235250.1 - 1 - 2 307 76
K01930.1 - 1 - 2 232 76
L01122.1 - 1 - 2 260 76

View Full Product Details