The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021835.3 | NP_068607.1 | 1 - 1 | 1644 | 106 |
GenBank mRNA | BC078738.1 | - | 1 - 1 | 881 | 106 |
X17163.1 | - | 1 - 1 | 106 |