Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BDH1
Entrez Gene ID
851239
Gene Name
Gene Aliases
YAL060W
Chromosome Location
Chr.I: 35155 - 36303 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178202.2 NP_009341.2 1 - 1 1053 82

View Full Product Details