Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMP23
Entrez Gene ID
852336
Gene Name
Gene Aliases
YBR047W
Chromosome Location
Chr.II: 331833 - 332360 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178395.1 NP_009603.1 1 - 1 449 99

View Full Product Details