Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMY2
Entrez Gene ID
852470
Gene Name
Gene Aliases
YBR172C
Chromosome Location
Chr.II: 579150 - 581372 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178520.3 NP_009731.4 1 - 1 2147 84

View Full Product Details