Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ILV6
Entrez Gene ID
850348
Gene Name
Gene Aliases
YCL009C
Chromosome Location
Chr.III: 104619 - 105548 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178658.1 NP_009918.1 1 - 1 850 69

View Full Product Details