Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEU2
Entrez Gene ID
850342
Gene Name
Gene Aliases
YCL018W
Chromosome Location
Chr.III: 91324 - 92418 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178665.1 NP_009911.2 1 - 1 946 85

View Full Product Details