Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GID7
Entrez Gene ID
850318
Gene Name
Gene Aliases
MOH2, YCL039W
Chromosome Location
Chr.III: 52645 - 54882 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178684.1 NP_009891.1 1 - 1 2155 95

View Full Product Details