Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRD1
Entrez Gene ID
850301
Gene Name
Gene Aliases
YCL057W
Chromosome Location
Chr.III: 24768 - 26906 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178701.1 NP_009874.1 1 - 1 2051 103

View Full Product Details