Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRP43
Entrez Gene ID
850401
Gene Name
Gene Aliases
YCR035C
Chromosome Location
Chr.III: 191834 - 193018 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178749.1 NP_009964.2 1 - 1 1052 99

View Full Product Details