Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SED4
Entrez Gene ID
850431
Gene Name
Gene Aliases
YCR067C
Chromosome Location
Chr.III: 233125 - 236322 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178778.1 NP_009993.1 1 - 1 3056 77

View Full Product Details