Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APC11
Entrez Gene ID
851554
Gene Name
Gene Aliases
YDL008W
Chromosome Location
Chr.IV: 433497 - 433994 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180067.3 NP_010276.3 1 - 1 341 98

View Full Product Details