Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERP3
Entrez Gene ID
851544
Gene Name
Gene Aliases
YDL018C
Chromosome Location
Chr.IV: 422833 - 423510 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180077.1 NP_010266.1 1 - 1 457 86

View Full Product Details