Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAA1
Entrez Gene ID
851643
Gene Name
Gene Aliases
YDR071C
Chromosome Location
Chr.IV: 588827 - 589402 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180379.1 NP_010356.1 1 - 1 155 93

View Full Product Details