Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSM24
Entrez Gene ID
851755
Gene Name
Gene Aliases
YDR175C, mS35
Chromosome Location
Chr.IV: 813193 - 814152 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180482.1 NP_010460.1 1 - 1 753 71

View Full Product Details