Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAD4
Entrez Gene ID
851919
Gene Name
Gene Aliases
HSK2, YDR320C-A
Chromosome Location
Chr.IV: 1108280 - 1108498 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184470.3 NP_710144.3 1 - 1 147 97

View Full Product Details