Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLG1
Entrez Gene ID
852079
Gene Name
Gene Aliases
YDR468C
Chromosome Location
Chr.IV: 1398026 - 1398700 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180776.3 NP_010756.3 1 - 1 549 60

View Full Product Details