Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCN4
Entrez Gene ID
856709
Gene Name
Gene Aliases
AAS101, AAS3, ARG9, YEL009C
Chromosome Location
Chr.V: 138918 - 139763 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178824.3 NP_010907.3 1 - 1 652 61

View Full Product Details