Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMD6
Entrez Gene ID
856637
Gene Name
Gene Aliases
YEL072W
Chromosome Location
Chr.V: 13720 - 14415 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178887.1 NP_010842.1 1 - 1 350 73

View Full Product Details