Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHO1
Entrez Gene ID
856748
Gene Name
Gene Aliases
PSS1, YER026C
Chromosome Location
Chr.V: 207644 - 208474 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178917.3 NP_010943.3 1 - 1 647 96

View Full Product Details