Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HVG1
- Entrez Gene ID
-
856761
- Gene Name
- Gene Aliases
-
YEM9, YER039C
- Chromosome Location
-
Chr.V: 228456 - 229205 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001178930.1 |
NP_010956.1 |
1 - 1 |
452 |
92 |
- Gene Symbol
- FMP49
- Entrez Gene ID
-
20098144
- Gene Name
- Gene Aliases
-
YER038W-A
- Chromosome Location
-
Chr.V: 228451 - 228831 on Build R64
View Full Product Details