Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BOI2
Entrez Gene ID
856850
Gene Name
Gene Aliases
BEB1, YER114C
Chromosome Location
Chr.V: 390590 - 393712 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179004.1 NP_011039.1 1 - 1 3044 73

View Full Product Details