Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IOC3
Entrez Gene ID
850567
Gene Name
Gene Aliases
YFR013W
Chromosome Location
Chr.VI: 169922 - 172285 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179978.1 NP_116668.1 1 - 1 2054 88

View Full Product Details