Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGD1
Entrez Gene ID
852860
Gene Name
Gene Aliases
HRS1, MED3, YGL025C
Chromosome Location
Chr.VII: 448764 - 449957 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180890.1 NP_011490.2 1 - 1 647 68

View Full Product Details