Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNC1
Entrez Gene ID
852846
Gene Name
Gene Aliases
YGL037C
Chromosome Location
Chr.VII: 427297 - 427947 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180902.3 NP_011478.3 1 - 1 546 66

View Full Product Details