Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VID30
Entrez Gene ID
852624
Gene Name
Gene Aliases
GID1, YGL227W
Chromosome Location
Chr.VII: 69671 - 72547 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181093.1 NP_011287.1 1 - 1 2246 77

View Full Product Details