Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRP38
Entrez Gene ID
852966
Gene Name
Gene Aliases
YGR075C
Chromosome Location
Chr.VII: 636147 - 636875 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181204.3 NP_011589.3 1 - 1 642 99

View Full Product Details