Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASK10
Entrez Gene ID
852989
Gene Name
Gene Aliases
RGC2, YGR097W
Chromosome Location
Chr.VII: 678695 - 682135 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181226.1 NP_011611.1 1 - 1 3249 72

View Full Product Details