Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADE3
Entrez Gene ID
853118
Gene Name
Gene Aliases
YGR204W
Chromosome Location
Chr.VII: 905934 - 908774 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181333.3 NP_011720.3 1 - 1 2750 78

View Full Product Details