Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EFG1
Entrez Gene ID
853188
Gene Name
Gene Aliases
YGR271C-A, YGR272C
Chromosome Location
Chr.VII: 1037800 - 1038501 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184474.3 NP_076891.4 1 - 1 645 77

View Full Product Details