Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAM35
Entrez Gene ID
856483
Gene Name
Gene Aliases
FMP20, TOB38, TOM38, YHR083W
Chromosome Location
Chr.VIII: 272627 - 273616 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179213.1 NP_011951.1 1 - 1 649 84

View Full Product Details