Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTM1
Entrez Gene ID
856509
Gene Name
Gene Aliases
YHR109W
Chromosome Location
Chr.VIII: 330310 - 332067 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179239.1 NP_011977.1 1 - 1 1647 102

View Full Product Details