Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNL1
Entrez Gene ID
854796
Gene Name
Gene Aliases
YIL016W
Chromosome Location
Chr.IX: 321454 - 321933 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179366.1 NP_012248.1 1 - 1 350 79

View Full Product Details