Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SUC2
Entrez Gene ID
854644
Gene Name
Gene Aliases
YIL162W
Chromosome Location
Chr.IX: 37385 - 38983 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179510.1 NP_012104.1 1 - 1 1440 94

View Full Product Details