Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYS1
Entrez Gene ID
854852
Gene Name
Gene Aliases
YIR034C
Chromosome Location
Chr.IX: 419615 - 420736 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179556.3 NP_012300.3 1 - 1 1048 109

View Full Product Details