Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTK2
Entrez Gene ID
853450
Gene Name
Gene Aliases
YJL006C
Chromosome Location
Chr.X: 423138 - 424109 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181440.1 NP_012528.1 1 - 1 852 118

View Full Product Details